Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Flt-3/Flk-2 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7681U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7681U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7681U100 Supplier R&D Systems Supplier No. FAB7681U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Flt-3/Flk-2/CD135
Applications Flow Cytometry
Classification Monoclonal
Clone 113308
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q00342
Gene Alias CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, Flk-2, FLK2, Flt3, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Gene Symbols FLT3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Flt-3/Flk-2 Asn28-Ser544 (predicted) Accession # Q00342
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2322
Test Specificity Detects mouse Flt-3/Flk-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Flt-3 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.