Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Flt-3/Flk-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | Flt-3/Flk-2/CD135 |
---|---|
Clone | 113315 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Mouse samples. It is validated for Western Blot.Specifications
Flt-3/Flk-2/CD135 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, Flk-2, FLK2, Flt3, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
FLT3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
113315 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q00342 | |
2322 | |
Mouse myeloma cell line NS0-derived recombinant mouse Flt-3/Flk-2 Asn28-Ser544 Accession # Q00342 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Flt-3/Flk-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) Flt-3 is observed and no cross-reactivity with rhPDGF-R alpha , rhPDGF-R beta , or rhSCF-R is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title