Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Flt-3 Ligand (FLT3L) Recombinant Protein

Catalog No. PIRP87871
Encompass
Click to view available options
Quantity:
10 μg
100 μg
25 μg

CHO cells Recombinant Protein

FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49772
Concentration 200 μg/mL
For Use With (Application) Control, ELISA, Western Blot
Formulation PBS with no preservative; pH 6
Gene ID (Entrez) 14256
Molecular Weight (g/mol) 18.3 kD
Name Mouse FLT3LG (aa 27-188)
Quantity 100 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration 0.01 ng/ μg cytokine
Gene Alias FL; Flt 3 ligand; Flt3 L; Flt3 ligand; Flt3l; Flt3lg; fms related receptor tyrosine kinase 3 ligand; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine
Gene Symbol Flt3l
Specific Reactivity Mouse
Conjugate Unconjugated
Recombinant Recombinant
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.