Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Follistatin-like 1/FSTL1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $643.50
Specifications
Antigen | Follistatin-like 1/FSTL1 |
---|---|
Clone | 228210 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Follistatin-like 1/FSTL1 Monoclonal specifically detects Follistatin-like 1/FSTL1 in Mouse samples. It is validated for Western Blot.Specifications
Follistatin-like 1/FSTL1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q62356 | |
11167 | |
Mouse myeloma cell line NS0-derived recombinant mouse Follistatin-like 1/FSTL1 Glu19-Ile306 Accession # Q62356 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Follistatin-like 1/FSTL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human Follistatin-like 1/FSTL1 is observed. |
228210 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Flik, FLJ50214, follistatin-like 1, Follistatin-like protein 1, Follistatin-related Protein, follistatin-related protein 1, FRP, FRPFLJ52277, FSL1, FSTL1, TSC-36 | |
FSTL1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title