Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FoxD3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5090SP
This item is not returnable.
View return policy
Description
FoxD3 Polyclonal specifically detects FoxD3 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
FoxD3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AIS1, CWH3, forkhead box D3, forkhead box protein D3, Genesis, HFH2, HNF3/FH transcription factor genesis | |
Goat | |
Affinity Purified | |
RUO | |
27022 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q61060 | |
FOXD3 | |
E. coli-derived recombinant mouse FoxD3 Met1-Val130 Accession # Q61060 | |
25 μg | |
Primary | |
Detects mouse FoxD3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FoxD3, rhFoxB2, rhFoxH1, and recombinant mouse FoxC2 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction