Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FoxD3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB5090
This item is not returnable.
View return policy
Description
FoxD3 Monoclonal specifically detects FoxD3 in Mouse samples. It is validated for Immunocytochemistry.Specifications
FoxD3 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q61060 | |
FOXD3 | |
E. coli-derived recombinant mouse FoxD3 Met1-Val130 Accession # Q61060 | |
100 μg | |
Primary | |
Detects mouse FoxD3 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human FoxD3 is observed. | |
Mouse | |
Purified |
Immunocytochemistry | |
800602 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AIS1, CWH3, forkhead box D3, forkhead box protein D3, Genesis, HFH2, HNF3/FH transcription factor genesis | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27022 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction