Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FoxD3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5090SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5090SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5090SP Supplier R&D Systems Supplier No. MAB5090SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

FoxD3 Monoclonal specifically detects FoxD3 in Mouse samples. It is validated for Immunocytochemistry.

Specifications

Antigen FoxD3
Applications Immunocytochemistry
Classification Monoclonal
Clone 800602
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q61060
Gene Alias AIS1, CWH3, forkhead box D3, forkhead box protein D3, Genesis, HFH2, HNF3/FH transcription factor genesis
Gene Symbols FOXD3
Host Species Rat
Immunogen E. coli-derived recombinant mouse FoxD3 Met1-Val130 Accession # Q61060
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27022
Test Specificity Detects mouse FoxD3 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human FoxD3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.