Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FPRP/PTGFRN Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4495SP
This item is not returnable.
View return policy
Description
FPRP/PTGFRN Polyclonal specifically detects FPRP/PTGFRN in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
FPRP/PTGFRN | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD315, CD9P-1, FPRP, PTGFRN | |
Sheep | |
Affinity Purified | |
RUO | |
5738 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
NP_035327 | |
Ptgfrn | |
Mouse myeloma cell line NS0-derived recombinant mouse FPRP/PTGFRN, Arg22-Pro832, Accession # NP_035327 | |
25 μg | |
Primary | |
Detects mouse FPRP/PTGFRN in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction