Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FPRP/PTGFRN Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4495
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4495 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4495 Supplier R&D Systems Supplier No. BAF4495
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

FPRP/PTGFRN Polyclonal specifically detects FPRP/PTGFRN in Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen FPRP/PTGFRN
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells
Gene Accession No. NP_035327
Gene Alias CD315, CD9P-1, FPRP, PTGFRN
Gene Symbols Ptgfrn
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FPRP Arg22-Pro832 Accession # NP_035327
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5738
Test Specificity Detects mouse FPRP in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.