Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Free Hemoglobin ELISA Kit (Colorimetric)
SDP

Catalog No. NB599991KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB599991KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB599991KT Supplier Novus Biologicals™ Supplier No. NBP2599991KIT
Only null left
Add to Cart
Add to Cart

Quantification of target antigens specific to ANGPT1 antigen.

The Mouse Free Hemoglobin ELISA Kit (Colorimetric) measures mouse Free Hemoglobin in serum, plasma.

Specifications

Assay Range 0.781 to 50μg/mL
Assay Sensitivity 0.177μg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Application) ELISA
Gene Symbol ANGPT1
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Free Hemoglobin
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.