Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Frizzled-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.50
Specifications
Antigen | Frizzled-2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Frizzled-2 Polyclonal specifically detects Frizzled-2 in Mouse samples. It is validated for Western Blot.Specifications
Frizzled-2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, Frizzled2, fz-2, Fz2, FZD2, FzE2, hFz2 | |
FZD2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9JIP6 | |
2535 | |
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-2 Gln29-Leu168 Accession # Q9JIP6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Frizzled-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse (rm) Frizzled-1 is observed and less than 5% cross-reactivity with rmFrizzled-4, recombinant human Frizzled-5, rmFrizzled-6, and rmFrizzled-8 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title