Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Frizzled-7 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $2350.00
Specifications
Antigen | Frizzled-7 |
---|---|
Clone | 151107 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Frizzled-7 Monoclonal specifically detects Frizzled-7 in Mouse samples. It is validated for Western Blot.Specifications
Frizzled-7 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q61090 | |
8324 | |
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-7 Gln33-Leu185 (predicted) Accession # Q61090 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects recombinant mouse (rm) Frizzled-7 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-1, -2, -3, -4, -6 or -8 is observed. |
151107 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Frizzled7, fz-7, FZD7, FzE3frizzled-7, hFz7 | |
FZD7 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title