Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Frizzled-9 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $526.50
Specifications
Antigen | Frizzled-9 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
Frizzled-9 Polyclonal specifically detects Frizzled-9 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Frizzled-9 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CD349, CD349 antigen, frizzled (Drosophila) homolog 9, frizzled homolog 9 (Drosophila), frizzled-9, Frizzled9, fz-9, FZD3fzE6, FZD9, FzE6, hFz9 | |
FZD9 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Q9R216 | |
8326 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Frizzled-9 Leu24-Asp186 Accession # Q9R216 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Frizzled-9 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse (rm) Frizzled-2, rmFrizzled-3, rmFrizzled-4, rmFrizzled-6, rmFrizzled-7, and rmFrizzled-8 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title