Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse G-CSF Alexa Fluor 594-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems IC414T100UG
This item is not returnable.
View return policy
Description
G-CSF Monoclonal specifically detects G-CSF in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
G-CSF | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
E. coli-derived recombinant mouse G-CSF, Val31-Ala208, Accession No. P09920 | |
100 μg | |
1440.0 | |
Mouse | |
Supernatant |
Flow Cytometry | |
67604 | |
Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
P09920 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse G-CSF in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant human (rh) G-CSF, rhCNTF, rmIL-6, rmLIF, or rmOSM. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction