Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse G-CSF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB414100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB414100 100 μg
MAB414SP 25 μg
MAB414500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB414100 Supplier R&D Systems Supplier No. MAB414100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 32 publications

G-CSF Monoclonal specifically detects G-CSF in Mouse samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen G-CSF
Applications Western Blot, Neutralization, Flow Cytometry, ELISA
Classification Monoclonal
Clone 67604
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.01-0.03 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P09920
Gene Alias C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Gene Symbols CSF3
Host Species Rat
Immunogen E. coli-derived recombinant mouse G-CSF Val31-Ala208 Accession # P09920
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1440
Test Specificity Detects mouse G-CSF in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant human (rh) G-CSF, rhCNTF, rmIL-6, rmLIF, or rmOSM.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.