Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse G-CSF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB414RSP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB414R-SP 25 μg
MAB414R-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB414R-SP Supplier R&D Systems Supplier No. MAB414RSP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

G-CSF Monoclonal specifically detects G-CSF in Mouse samples. It is validated for ELISA.

Specifications

Antigen G-CSF
Applications ELISA
Classification Monoclonal
Clone 67604R
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. P09920
Gene Alias C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Host Species Rat
Immunogen E. coli-derived recombinant mouse G-CSF Accession # P09920
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 1440
Test Specificity Detects mouse G-CSF in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.