Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse G-CSFR/CD114 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. FAB6039G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6039G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6039G Supplier R&D Systems Supplier No. FAB6039G
This item is not returnable. View return policy

Sheep Polyclonal Antibody

G-CSFR/CD114 Polyclonal specifically detects G-CSFR/CD114 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen G-CSFR/CD114
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Gene Accession No. P40223
Gene Alias CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), CSF3R, Csfgr, G-CSF R, G-CSF receptor, GCSFR, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor
Gene Symbols Csf3r
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse G-CSF R/CD114 Cys26-Asp626 Accession # P40223
Purification Method Affinity Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1441
Test Specificity Detects mouse G-CSF R/CD114 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human GM-CSF R alpha and recombinant mouse GM-CSF R alpha is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.