Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Galectin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1245
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1245 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1245 Supplier R&D Systems Supplier No. MAB1245
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Galectin-1 Monoclonal specifically detects Galectin-1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Galectin-1
Applications Western Blot
Classification Monoclonal
Clone 201066
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P16045
Gene Alias 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, BHL, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, Galectin1, GBP, HBL, HLBP14, HPL, L-14, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, LGALS1, Putative MAPK-activating protein PM12, S-Lac lectin 1
Gene Symbols LGALS1
Host Species Rat
Immunogen E. coli-derived recombinant mouse Galectin-1 Ala2-Glu135 Accession # P16045
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3956
Test Specificity Detects mouse Galectin-1 in direct ELISAs and Western blots. In Western blots, this antibody shows 100% cross-reactivity with recombinant human (rh) Galectin-1 but no cross-reactivity with rhGalectin-2, -8, rmGalectin-3, -4, or -7.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.