Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Galectin-1 PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems IC1245P
This item is not returnable.
View return policy
Description
Galectin-1 Polyclonal specifically detects Galectin-1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Galectin-1 | |
Polyclonal | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, BHL, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, Galectin1, GBP, HBL, HLBP14, HPL, L-14, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, LGALS1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
Goat | |
Affinity Purified | |
RUO | |
3956 | |
Mouse | |
IgG |
Flow Cytometry | |
R-PE | |
P16045 | |
LGALS1 | |
E. coli-derived recombinant mouse Galectin-1 Ala2-Glu135 Accession # P16045 | |
100 Tests | |
Primary | |
Detects mouse Galectin-1 in ELISAs and Western blots. In sandwich ELISAs, less than 5% cross-reactivity with recombinant human (rh) Galectin-1 and less than 0.5% cross-reactivity with recombinant mouse (rm) Galectin-3, rhGalectin-4, rmGalectin-7, and rhGalectin-8 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction