Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Galectin-7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1304
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1304 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1304 Supplier R&D Systems Supplier No. MAB1304
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Galectin-7 Monoclonal specifically detects Galectin-7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Galectin-7
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 212923
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAK29385
Gene Alias Gal-7, GAL7, galectin 7, galectin-7, Galectin7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1
Gene Symbols LGALS7
Host Species Rat
Immunogen E. coli-derived recombinant mouse Galectin-7 Ser2-Phe136 Accession # AAK29385
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3963
Test Specificity Detects mouse Galectin-7 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Galectin-1, -3, -4, recombinant human (rh) Galectin-2, -7, or -8 is observed. In Western blots, approximately 20% cross-reactivity with rhGalectin-3, approximately 10% cross-reactivity with rhGalectin-7, and no cross-reactivity with rhGalectin-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.