Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Galectin-9 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3535
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3535 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3535 Supplier R&D Systems Supplier No. BAF3535
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Galectin-9 Polyclonal specifically detects Galectin-9 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Galectin-9
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O08573
Gene Alias ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Gene Symbols LGALS9
Host Species Goat
Immunogen E. coli-derived recombinant mouse Galectin-9 short isoform Ala2-Thr353; aa148-178 deleted Accession # O08573
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3965
Test Specificity Detects mouse Galectin-9 in Western blots. In this format, less than 1% cross-reactivity with recombinant human (rh) Galectin-2, rhGalectin-8, recombinant mouse (rm) Galectin-1, rmGalectin-3, rmGalectin-4, and rmGalectin-7 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.