Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Gamma Globulin, Invitrogen™

Catalog No. PI31878
Click to view available options
Quantity:
10 mg

Mouse Control

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Storage: After opening, dilute only enough antibody for a single day’s use. Country of Origin: USA

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody's species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4° C
Form Liquid
Product Type Mouse Gamma Globulin
Quantity 10 mg
Concentration 11.0 mg/mL
Formulation PBS with no preservative; pH 7.6
For Use With (Application) Control, ChIP Assay, Immunoprecipitation, Flow Cytometry
Isotype Ig
Regulatory Status RUO
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.