Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Gas6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB986
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB986 500 μg
MAB986SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB986 Supplier R&D Systems Supplier No. MAB986
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Gas6 Monoclonal specifically detects Gas6 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Gas6
Applications Western Blot
Classification Monoclonal
Clone 126618
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q61592.2
Gene Alias AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6
Gene Symbols GAS6
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Gas6 Asp115-Pro674 (Del Pro530) Accession # Q61592.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2621
Test Specificity Detects mouse Gas6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Gas6.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.