Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Gastrokine 1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7287
This item is not returnable.
View return policy
Description
Gastrokine 1 Monoclonal specifically detects Gastrokine 1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Gastrokine 1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
NP_079742 | |
GKN1 | |
E. coli-derived recombinant mouse Gastrokine 1 Tyr38-Tyr201 Accession # NP_079742 | |
100 μg | |
Primary | |
Detects mouse Gastrokine 1 in direct ELISAs and mouse and rat Gastrokine 1 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) Gastrokine 1 and no cross-reactivity with rhBlottin is observed. | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
759821 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
18 kDa antrum mucosa protein, AMP18, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, GKN1, Protein CA11 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
56287 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction