Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse GATA-4 ELISA Kit (Colorimetric)
SDP

Catalog No. NB112490 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB112490 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB112490 Supplier Novus Biologicals™ Supplier No. NBP300434
Add to Cart
Add to Cart

Mouse GATA-4 ELISA Kit (Colorimetric) measures GATA-4 in Serum, plasma, cell supernatant, and other biological fluids.

Specifications

Assay Range 0.31 to 20 ng/mL
Assay Sensitivity 0.19 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse GATA-4 in. samples. No significant cross-reactivity or interference between Mouse GATA-4 and analogues was observed.
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 2626
Gene Symbol GATA4
Quantity 1 Kit
Research Discipline Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
Target GATA-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.