Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GDF-1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF858
This item is not returnable.
View return policy
Description
GDF-1 Polyclonal specifically detects GDF-1 in Mouse samples. It is validated for Western Blot.Specifications
GDF-1 | |
Polyclonal | |
Biotin | |
P20863 | |
GDF1 | |
E. coli-derived recombinant mouse GDF-1 His238-Arg357 Accession # P20863 | |
50 μg | |
Primary | |
Detects mouse GDF-1 in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse (rm) GDF-3 is observed, less than 5% cross-reactivity with rmGDF-5 and rmGDF-6 is observed, and less than 1% cross-reactivity with rmGDF-8 and rmGDF-9 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
DORV, DTGA3, embryonic growth/differentiation factor 1, GDF1, GDF-1, growth differentiation factor 1 | |
Goat | |
Affinity Purified | |
RUO | |
2657 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction