Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GDF-15 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6385
This item is not returnable.
View return policy
Description
GDF-15 Polyclonal specifically detects GDF-15 in Mouse samples. It is validated for Immunohistochemistry.Specifications
GDF-15 | |
Polyclonal | |
Unconjugated | |
Q9Z0J7 | |
GDF15 | |
E. coli-derived recombinant mouse GDF-15 Ser189-Ala303 Accession # Q9Z0J7 | |
100 μg | |
Primary | |
Detects mouse GDF-15 in direct ELISAs. | |
Mouse | |
IgG |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
GDF15, growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDF, PDFGDF-15, PLAB, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGF-beta, PTGFBPTGF-beta | |
Sheep | |
Affinity Purified | |
RUO | |
9518 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction