Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse GDF-3 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF958
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF958 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF958 Supplier R&D Systems Supplier No. BAF958
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

GDF-3 Polyclonal specifically detects GDF-3 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen GDF-3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q07104
Gene Alias GDF3, GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6, Vgr-2
Gene Symbols GDF3
Host Species Goat
Immunogen E. coli-derived recombinant mouse GDF-3 Ile255-Gly366 Accession # Q07104
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9573
Test Specificity Detects mouse GDF-3 in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse (rm) GDF-1 is observed and less than 5% cross-reactivity with rmGDF-5, rmGDF-6, rmGDF-8 and rmGDF-9 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.