Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse GDF-5/BMP-14 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM853
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM853 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM853 Supplier R&D Systems Supplier No. BAM853
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

GDF-5/BMP-14 Monoclonal specifically detects GDF-5/BMP-14 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen GDF-5/BMP-14
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 90509
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. P43027
Gene Alias BMP14, BMP-14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF5, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2
Gene Symbols GDF5
Host Species Rat
Immunogen E. coli-derived recombinant mouse GDF-5/BMP-14 Ala376-Arg495 Accession # P43027
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8200
Test Specificity Detects mouse GDF-5/BMP-14 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.