Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse GDF-7/BMP-12 ELISA Kit (Colorimetric)
SDP

Catalog No. NB398038 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398038 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398038 Supplier Novus Biologicals™ Supplier No. NBP308139
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Supernatant
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 151449
Gene Symbol GDF7
Quantity 1 Kit
Research Discipline Cytokine Research
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym BMP12, GDF-7, growth differentiation factor 7, growth/differentiation factor 7
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.