Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GDF-9 Propeptide Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
Antigen | GDF-9 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
GDF-9 Polyclonal specifically detects GDF-9 in Mouse samples. It is validated for Western Blot.Specifications
GDF-9 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
GDF9 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
AAA53035 | |
2661 | |
Mouse myeloma cell line NS0-derived recombinant mouse GDF-9 Propeptide Glu30-Arg304 Accession # AAA53035 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse GDF-9 Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with mature recombinant mouse (rm) GDF-9, rmGDF-1 propeptide, and rmGDF-3 propeptide is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title