Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GFR alpha-4/GDNF R alpha-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | GFR alpha-4/GDNF R alpha-4/Persephin receptor |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Polyclonal specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Mouse samples. It is validated for Immunohistochemistry.Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
64096 | |
Mouse myeloma cell line NS0-derived recombinant mouse GFR alpha-4/GDNF R alpha-4 Asn24-Thr237 Accession # Q99JT2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse GFR alpha-4/GDNF R alpha-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human GFR alpha-4 is observed. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor | |
GFRA4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title