Learn More
Mouse GFR alpha-4/GDNF R alpha-4 Biotinylated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | GFR alpha-4/GDNF R alpha-4/Persephin receptor |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Biotin |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Gene Alias | GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor |
| Gene Symbols | GFRA4 |
| Host Species | Goat |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse GFR alpha-4/GDNF R alpha-4 aa 35-242 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.