Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GFR alpha-like Isoform A Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF5728
This item is not returnable.
View return policy
Description
GFR alpha-like Polyclonal specifically detects GFR alpha-like in Mouse samples. It is validated for Western Blot.Specifications
GFR alpha-like | |
Polyclonal | |
Biotin | |
Q6SJE0 | |
Sheep | |
Affinity Purified | |
RUO | |
404194 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot | |
Gfral | |
Chinese hamster ovary cell line CHO-derived recombinant mouse GFR alpha-like isoform A Gln20-Gly349 Accession # Q6SJE0 | |
50 μg | |
Primary | |
Detects mouse GFR alpha-like in Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse (rm) GFR alpha-3 and rmGFR alpha-4 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction