Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse GITR Ligand/TNFSF18 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB21772SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB21772SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB21772SP Supplier R&D Systems Supplier No. MAB21772SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen GITR Ligand/TNFSF18
Applications Western Blot
Classification Monoclonal
Clone 721926
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_899247
Gene Alias Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Gene Symbols TNFSF18
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18 Thr47-Ser173 Accession # NP_899247
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8995
Test Specificity Detects mouse GITR Ligand/TNFSF18 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) APRIL, rmBAFF, rmEDA, rmFas Ligand, rmLIGHT, rmOX40 Ligand, rmTNF-alpha , rmTRAIL, rmTRANCE, rmTWEAK, rm4-1BB Ligand, recombinant human (rh) EDA-A2, rhLTa1/b2, rhLTa2/b1, rhVEGI, or rhGITR Ligand is observed. In Western blots, no cross-reactivity with rm4-1BB, rmEDA, rmLIGHT, rmTRAIL, rhGITR Ligand, rhTNF-alpha , recombinant canine TNF-alpha , recombinant guinea pig TNF-alpha , or recombinant porcine TNF-alpha is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.