Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GITR Ligand/TNFSF18 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | GITR Ligand/TNFSF18 |
---|---|
Clone | 994529 |
Dilution | Flow Cytometry 0.25 ug/106 cells, Neutralization 1.2-12 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, Neutralization, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB21773SP
![]() |
R&D Systems
MAB21773SP |
25 μg |
Each for $200.50
|
|
|||||
MAB21773100
![]() |
R&D Systems
MAB21773100 |
100 μg |
Each for $456.00
|
|
|||||
Description
GITR Ligand/TNFSF18 Monoclonal antibody specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry, Neutralization, CyTOFSpecifications
GITR Ligand/TNFSF18 | |
Flow Cytometry 0.25 ug/106 cells, Neutralization 1.2-12 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18, Thr47-Ser173, Accession # Q7TS55 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
994529 | |
Flow Cytometry, Neutralization, CyTOF | |
Unconjugated | |
Rat | |
Mouse | |
Q7TS55 | |
8995 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title