Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse GITR Ligand/TNFSF18 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2177
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2177 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2177 Supplier R&D Systems Supplier No. BAF2177
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

GITR Ligand/TNFSF18 Polyclonal specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for ELISA Detection (Matched Antibody Pair).

Specifications

Antigen GITR Ligand/TNFSF18
Applications ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. Q80YG2
Gene Alias Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Gene Symbols TNFSF18
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18 (R&D Systems, Catalog # 2177-GL) Thr47-Ser173 Accession # Q80YG2
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8995
Test Specificity Detects mouse GITR Ligand/TNFSF18 in ELISAs. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human GITR Ligand is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.