Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GITR/TNFRSF18 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | GITR/TNFRSF18 |
---|---|
Clone | 2375B |
Dilution | Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg;/mL, Immunocytochemistry 8-25 μg;/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB52412SP
![]() |
R&D Systems
MAB52412SP |
25 μg |
Each for $200.50
|
|
|||||
MAB52412100
![]() |
R&D Systems
MAB52412100 |
100 μg |
Each for $491.00
|
|
|||||
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
GITR/TNFRSF18 | |
Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg;/mL, Immunocytochemistry 8-25 μg;/mL, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Mouse | |
O35714 | |
8784.0 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2375B | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Activation-inducible TNFR family receptor, AITR, AITRTNF receptor superfamily activation-inducible protein, CD357, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, TNFRSF18, tumor necrosis factor receptor superfamily, member 18 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse GITR/TNFRSF18, Accession No. O35714 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse GITR/TNFRSF18 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title