Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse GITR/TNFRSF18 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5241A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5241A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5241A Supplier R&D Systems Supplier No. FAB5241A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen GITR/TNFRSF18
Applications Flow Cytometry
Classification Monoclonal
Clone 108619
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O35714
Gene Alias Activation-inducible TNFR family receptor, AITR, AITRTNF receptor superfamily activation-inducible protein, CD357, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, TNFRSF18, tumor necrosis factor receptor superfamily, member 18
Gene Symbols TNFRSF18
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GITR/TNFRSF18 Met1-His153 Accession # O35714
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8784
Test Specificity Detects mouse GITR/TNFRSF18 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) 4-1BB, recombinant mouse (rm) CD27, rmCD30, rmEDAR, rmFas, rhGITR, rhHVEM, rmRANK, rhTROY, and rmTNF R1 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.