Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse GLP1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB170628
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 0.16-10 ng/mL
Assay Sensitivity 0.1 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse GLP1 in samples. No significant cross-reactivity or interference between Mouse GLP1 and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 2641
Gene Symbol GCG
Quantity 1 Kit
Research Discipline Cancer, Signal Transduction, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP
Target Mouse
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.