Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Glut2 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1440A100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1440A100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1440A100 Supplier R&D Systems Supplier No. FAB1440A100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Glut2 Monoclonal specifically detects Glut2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Glut2
Applications Flow Cytometry
Classification Monoclonal
Clone 205115
Conjugate APC
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and sodium azide
Gene Accession No. P14246
Gene Alias GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2
Gene Symbols SLC2A2
Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with mouse Glut2, Met1-Val523, Accession No. P14246
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6514.0
Test Specificity Detects mouse Glut2. Recognizes mouse Glut2-transfected NS0 cells but not NS0 control transfectants. It also detects Glut2 on mouse insulinoma beta TC-6 cells (1).
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.