Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Glypican 5 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
Antigen | Glypican 5 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Glypican 5 Polyclonal specifically detects Glypican 5 in Mouse samples. It is validated for Western Blot.Specifications
Glypican 5 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
bA93M14.1, glypican 5, glypican proteoglycan 5, glypican-5, GPC5 | |
GPC5 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q8CAL5 | |
2262 | |
Mouse myeloma cell line NS0-derived recombinant mouse Glypican 5 Gly26-Gly549 Accession # Q8CAL5 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Glypican 5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant human (rh) Glypican 5 is observed and less than 1% cross-reactivity with recombinant mouse Glypican 2, rhGlypican 3, and rhGlypican 6 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title