Learn More
Mouse GM-CSF Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 4 publications
Supplier:  R&D Systems AF415NA
Catalog No. AF415NA
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
GM-CSF Polyclonal specifically detects GM-CSF in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
Specifications
GM-CSF
Polyclonal
Unconjugated
Q14AD9
CSF2
E. coli-derived recombinant mouse GM-CSF Ala18-Lys141 Accession # Q14AD9
100 μg
Primary
Detects mouse GM-CSF in direct ELISAs and Western blots.
Mouse
IgG
Western Blot, Neutralization
LYOPH
Western Blot 0.1 ug/mL, Neutralization 0.01-0.03 ug/mL
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Goat
Affinity Purified
RUO
1437
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.