Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GM-CSF Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 12 publications
$227.50 - $2110.00
Specifications
Antigen | GM-CSF |
---|---|
Clone | MP122E9 |
Dilution | Western Blot 1 ug/mL, Neutralization 0.03-0.09 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, Neutralization, ELISA |
Classification | Monoclonal |
Description
GM-CSF Monoclonal specifically detects GM-CSF in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
GM-CSF | |
Western Blot 1 ug/mL, Neutralization 0.03-0.09 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
P01587 | |
1437 | |
Yeast derived recombinant mouse GM-CSF Accession # P01587 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse GM-CSF in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human GM-CSF. |
MP122E9 | |
Western Blot, Neutralization, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
CSF2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title