Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GM-CSF Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF415
This item is not returnable.
View return policy
Description
GM-CSF Polyclonal specifically detects GM-CSF in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
GM-CSF | |
Polyclonal | |
Biotin | |
Q14AD9 | |
CSF2 | |
E. coli-derived recombinant mouse GM-CSF (R&D Systems, Catalog # 415-ML) Ala18-Lys141 Accession # Q14AD9 | |
50 μg | |
Primary | |
Detects mouse GM-CSF in ELISAs and Western blots. In sandwich immunoassays, less than 0.02% cross-reactivity with recombinant human (rh) GM-CSF, rhM-CSF, recombinant mouse (rm) IL-4, rhIL-5, recombinant rat (rr) GM-CSF, rmM-CSF, recombinant porcine (rp) IL-4, rmIL-5, rpGM-CSF, rhIL-3, rrIL-4, rhIL-5 sR alpha , rhG-CSF, rmIL-3, rhIL-4 sR, rmG-CSF, rhIL-3 sR alpha , rmIL-4 R Fc Chimera, rhM-CSF R Fc Chimera, and rhIL-4 is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
Goat | |
Affinity Purified | |
RUO | |
1437 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction