Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse GM-CSF Quantikine ELISA Kit

Catalog No. p-4753897
Click to view available options
Quantity:
2 Plates
6 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Mouse GM-CSF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse GM-CSF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse GM-CSF in cell culture supernates (50 ul), serum (50 ul).

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 5.8pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and recombinant mouse GM-CSF
Sufficient For 2 x 96-well Plates
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 12981
Quantity 2 Plates
Sample Volume Cell Culture Supernates (50uL),Serum (50uL)
Synonym colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.