Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse GPVI Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6758
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6758 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6758 Supplier R&D Systems Supplier No. MAB6758
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

GPVI Monoclonal specifically detects GPVI in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen GPVI
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 784808
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P0C191
Gene Alias Glycoprotein 6, glycoprotein VI (platelet), GP6, GPIV, GPVIplatelet collagen receptor, MGC138168, platelet glycoprotein VI
Gene Symbols GP6
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GPVI Gly24-Lys265 Accession # P0C191
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51206
Test Specificity Detects mouse GPVI in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human GPVI is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.