Learn More
Mouse Gr-1/Ly-6G APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 5 publications
Supplier:  R&D Systems FAB1037A100
 View more versions of this product
Catalog No. FAB1037A100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Gr-1/Ly-6G
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Ly6g
The immunogen for this antibody is Ly-6G (Gr-1, Gr1).
100 Tests
Primary
Detects mouse Gr-1/Ly-6G. Weak cross-reactivity with Ly-6C is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b
Flow Cytometry
RB6-8C5
Flow Cytometry 10 uL/10^6 cells
C6orf23, G6D, Ly-6C, LY6-D, Ly6G, Ly-6G, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
58530
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.