Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Mouse Granzyme B Platinum ELISA Kit Non-distribution product as customer accommodation.

For the quantitative detection of mouse Granzyme B

Manufacturer:  Invitrogen™ BMS6029

 View more versions of this product

Catalog No. 50-112-5445


Add to cart

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse Granzyme B

Description

Description

Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.
Specifications

Specifications

P04187
Biotin
14939
Adhesive Plate Covers
96 tests
1.6 pg/mL
50μL
Mouse
1 hr 20 min
Mouse
Granzyme 2, Fragmentin 2, Human lymphocyte protein
3.5%
7.8-500 pg/mL
ELISA
Colorimetric Microplate Reader
ELISA Kit
RUO
Plasma,Serum,Supernatant
Plasma,50 μL|Serum,50 μL|Supernatant,50 μL
2-8°C
3 hr 30 min
Gzmb
4.1%
HRP
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only