Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Granzyme G Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
Antigen | Granzyme G |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Granzyme G Polyclonal specifically detects Granzyme G in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Granzyme G | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
Ctla7, Ctla-7, GrzG, GZMG | |
GZMG | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
P13366 | |
14944 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Granzyme G Ile21-Leu248 Accession # P13366 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Granzyme G in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse Granzyme D is observed and less than 5% cross-reactivity with recombinant human Granzyme H and recombinant mouse Granzyme B is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title